Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF64704685

Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3

sphere, 1.0mm diameter, 2.50μm sphericity, precision sphere, grade 100, 100pcs

Synonym(s):

FE246810

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

spheres (grade 100)

quality

precision sphere

packaging

1 ea of 100 pieces

manufacturer/tradename

Goodfellow 647-046-85

diam.

1.0 mm

sphericity

2.50 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service