Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF62304476

Graphene dispersion

solids content 40%,100 g, viscosity 5.5 Pa.s

Synonym(s):

GK00NK10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiCxHyOz
UNSPSC Code:
12141908
NACRES:
NA.23

Product Name

Graphene, ink, solids content 40%, 100 g, viscosity 5.5 Pa.s 

form

solid

manufacturer/tradename

Goodfellow 623-044-76

W

100 g

viscosity

5.5 Pa.s

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service