Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF49084498

Lutetium

lump, 25 mm max. lump size, weight 2 g, purity 99.9%

Synonym(s):

LU006100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Lu
CAS Number:
Molecular Weight:
174.97
MDL number:
UNSPSC Code:
12141608
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

lumps

manufacturer/tradename

Goodfellow 490-844-98

resistivity

54 μΩ-cm, 20°C

weight

2 g

max. lump size

25 mm

bp

3402 °C (lit.)

mp

1663 °C (lit.)

density

9.84 g/mL at 25 °C (lit.)

SMILES string

[Lu]

InChI

1S/Lu

InChI key

OHSVLFRHMCKCQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service