Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF48727547

Inconel® 625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5

foil, 100 x 100mm, 0.2mm thickness, annealed

Synonym(s):

NI040240

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (annealed)

manufacturer/tradename

Goodfellow 487-275-47

size × thickness

100 mm × 100 mm × 0.2 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service