Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF46146225

Zinc oxide

powder, max. particle size 45 micron, weight 50 g, high purity 99.999%

Synonym(s):

ZN526010

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352303
NACRES:
NA.23

Assay

99.999%

form

powder

manufacturer/tradename

Goodfellow 461-462-25

weight

50 g

max. part. size

45 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service