Skip to Content
MilliporeSigma
All Photos(2)

Documents

GF45937794

Zinc

foil, 50mm disks, thickness 0.5mm, as rolled, 99.99+%

Synonym(s):

Zinc, ZN000315

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 459-377-94

resistivity

5.8 μΩ-cm, 20°C

diam. × thickness

50 mm × 0.5 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuanhui Xia et al.
Journal of nanoscience and nanotechnology, 11(12), 10506-10510 (2012-03-14)
We have investigated the properties of Mn-doped ZnO nanocrystalline film growing on zinc foil by the hydrothermal method. X-ray photoelectron spectroscopy shows that the manganese ions exist as Mn2+ in the film. From UV-vis spectra, we observe a red shift
Tandra Ghoshal et al.
Nanotechnology, 19(6), 065606-065606 (2008-02-13)
ZnO nanowire arrays were synthesized on zinc foil by a simple thermal evaporation process at relatively low temperature. Morphology and size controlled synthesis of the ZnO nanostructures was achieved by variation of the synthesis temperature, reaction time and the surface
Conghua Lu et al.
Chemical communications (Cambridge, England), (33)(33), 3551-3553 (2006-08-22)
Large-scale, ultralong ZnO nanowire and nanobelt arrays with honeycomb-like micropatterns have been fabricated by hydrothermal oxidation of zinc foil in aqueous alkaline (NH4)2S2O8 solutions.
Lance Brockway et al.
Physical chemistry chemical physics : PCCP, 15(17), 6260-6267 (2013-03-23)
A simple method for the large-scale synthesis of gram quantities of compound semiconductor nanowires without the need for any external catalysts or templates is presented. This method is demonstrated using zinc phosphide (Zn3P2) and zinc antimonide (β-Zn4Sb3) nanowires as example
Rui Gao et al.
Nanoscale, 5(5), 1894-1901 (2013-01-29)
In this paper, ZnO nanorod-nanosheet hierarchical structures were fabricated using a facile method on zinc foil and used as flexible photoanodes in dye-sensitized solar cells (DSCs). Compared to nanorods (NRs) obtained by the dissolution-precipitation method, the nanorod-nanosheet (NR-NS) hierarchical structures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service