GF44759054
Roses metal alloy, Bi50Pb28Sn22
100g lump, 100mm max. lump size
Synonym(s):
BI036100
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
lumps
manufacturer/tradename
Goodfellow 447-590-54
weight
100 g
max. lump size
100 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service