Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF42099981

Rubidium

lump, 5 mm max. lump size, weight 1 g, purity 99.9%

Synonym(s):

RB006100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141805
NACRES:
NA.23

Assay

99.9%

form

lumps

manufacturer/tradename

Goodfellow 420-999-81

weight

1 g

max. lump size

5 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service