Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF37976147

Silicon carbide

fiber, tex number 209, length 5 m, filament diameter 0.015mm

Synonym(s):

SI675721

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101521
NACRES:
NA.23

form

fibers

manufacturer/tradename

Goodfellow 379-761-47

L

5 m

diam.

0.015 mm , filament diameter

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service