Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF26319944

Stainless Steel - AISI 302 alloy, FeCr18Ni8

50m insulated wire, 0.025mm conductor diameter, 0.002mm insulation thickness, 2 micron polyesterimide insulation

Synonym(s):

FE205811

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

2 micron polyesterimide insulation

form

wire (insulated)

manufacturer/tradename

Goodfellow 263-199-44

L

50 m

conductor diam.

0.025 mm

insulation thickness

0.002 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service