Skip to Content
MilliporeSigma
All Photos(2)

Documents

GF17475903

Nickel

rod, 200mm, diameter 6.35mm, 99%

Synonym(s):

Nickel, NI007955

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 174-759-03

resistivity

6.97 μΩ-cm, 20°C

L × diam.

200 mm × 6.35 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal Cox et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 505-513 (2014-04-08)
Nickel-titanium closed-coil springs are purported to deliver constant forces over extended ranges of activation and working times. In-vivo studies supporting this claim are limited. The objective of this study was to evaluate changes in force-decay properties of nickel-titanium closed-coil springs
Gonensin O Bozdag et al.
Gene, 538(2), 251-257 (2014-02-04)
Nickel is an essential micronutrient due to its involvement in many enzymatic reactions as a cofactor. However, excess of this element is toxic to biological systems. Here, we constructed a cDNA library from Beta maritima and screened it in the
Egor Geist et al.
Natural product reports, 31(4), 441-448 (2014-02-28)
This Highlight covers the current status of relatively unexplored sp(3)-sp(3) cross-coupling reactions with particular focus on natural product and related syntheses.
Giovanna Visioli et al.
Bulletin of environmental contamination and toxicology, 92(4), 490-496 (2013-11-30)
In vitro short-term chronic phytotoxicity germination and root elongation test were applied to test the effects of nickel (Ni) in seed germination and root elongation in six plants species: Cucumis sativus (Cucurbitaceae), Lepidium sativum and Brassica nigra (Brassicaceae), Trifolium alexandrinum
Daniel H Rothman et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(15), 5462-5467 (2014-04-08)
The end-Permian extinction is associated with a mysterious disruption to Earth's carbon cycle. Here we identify causal mechanisms via three observations. First, we show that geochemical signals indicate superexponential growth of the marine inorganic carbon reservoir, coincident with the extinction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service