Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF12231223

Gold

microleaf disks, 10mm, thinness 0.25μm, specific density 483μg/cm2, removable support, 99.99+%

Synonym(s):

Gold, AU004925

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 122-312-23

resistivity

2.05 μΩ-cm, 0°C

diam. × thickness

10 mm × 0.25 μm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingbo Zu et al.
Journal of biomedical nanotechnology, 10(6), 982-992 (2014-04-23)
Electroporation figured prominently as an effective nonviral gene delivery approach for its balance on the transfection efficiency and cell viability, no restrictions of probe or cell type, and operation simplicity. The commercial electroporation systems have been widely adopted in the
Masaki Ujihara et al.
Journal of nanoscience and nanotechnology, 14(7), 4906-4910 (2014-04-25)
Effect of structure and size of Au nanoparticles (AuNPs) on fluorescence behavior of uranine was examined. Confeito-like AuNPs with different sizes (30 nm, 60 nm and 100 nm, respectively) had plasmon absorption bands at 555, 600 and 660 nm, while
Maria Jose Santos-Martinez et al.
Journal of biomedical nanotechnology, 10(6), 1004-1015 (2014-04-23)
The increasing use of gold nanoparticles in medical diagnosis and treatment has raised the concern over their blood compatibility. The interactions of nanoparticles with blood components may lead to platelet aggregation and endothelial dysfunction. Therefore, medical applications of gold nanoparticles
Lin Luo et al.
Journal of nanoscience and nanotechnology, 14(6), 4036-4042 (2014-04-18)
A novel immunosensor of human immune globulin (IgG) was fabricated based on the fluorescence transfer between luminescent semiconductor quantum dots (QDs) and gold nanoparticles (AuNPs). AuNPs and CdSe/ZnS QDs were respectively labeled with immune reaction pair:IgG and goat anti-human immunoglobulin
Jiajing Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4124-4138 (2014-04-18)
Gold nanostructures with promising applications in biomedical field have attracted great attention. However, some fundamental questions other than the development of novel applications should be elucidated before they can actually serve as biomedicines in the clinic. Bio-safety is one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service