Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF00044682

Ytterbium

foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%

Synonym(s):

Ytterbium, YB000205

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12141616
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

foil

manufacturer/tradename

Goodfellow 000-446-82

resistivity

28 μΩ-cm, 20°C

L × W × thickness

25 mm × 25 mm × 0.005 mm

bp

1196 °C (lit.)

mp

824 °C (lit.)

density

6.54 g/mL at 25 °C (lit.)

SMILES string

[Yb]

InChI

1S/Yb

InChI key

NAWDYIZEMPQZHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ru Zhou et al.
Journal of nanoscience and nanotechnology, 14(6), 4196-4200 (2014-04-18)
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission
Wanyue Shao et al.
Journal of nanoscience and nanotechnology, 14(5), 3887-3892 (2014-04-17)
Water-soluble BaYF5:Yb3+/Er3+ nanoparticles with the surface functionalized by a layer of poly(acrylic acid) (PAA) were synthesized via a facile one-step PAA-assisted hydrothermal method. Bovine serum albumin (BSA) protein was conjugated with BaYF5:Yb3+/Er3+ upconversion nanoparticles via free carboxylic acid groups on
Sethuraman Gayathri et al.
Carbohydrate polymers, 121, 302-308 (2015-02-11)
In this study, chitosan functionalized LaF3:Yb,Er upconverting nanotransducers (UCNTs) with controlled size and shape have been successfully synthesized by a facile one pot precipitation method. The chitosan encapsulated UCNTs show bright upconversion fluorescence upon excitation with 974 nm NIR region.
Ying Xie et al.
Journal of nanoscience and nanotechnology, 14(6), 4486-4489 (2014-04-18)
Eu3+ and Yb3+ codoped Y2O3 phosphors were synthesized by the sol-gel method. The phosphors possess absorption in the region of 300-550 nm, exhibiting an intense NIR emission of Yb3+ around 1000 nm, which is suitable for matching the maximum spectral
Thorben Rinkel et al.
Nanoscale, 6(23), 14523-14530 (2014-10-28)
We have studied the growth behaviour of sub-10 nm NaYF₄ upconversion nanocrystals of the hexagonal β-phase and the cubic α-phase. Ostwald-ripening of such particles in oleic acid/octadecene solvent results in broadening of the particle size distribution if the colloid contains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service