Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

E32754

Sigma-Aldrich

4,6-O-Ethylidene-α-D-glucose

Synonym(s):

4,6-O-Ethylidene α-D-glucopyranose, Ethylidene glucose, NSC 89726

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O6
CAS Number:
Molecular Weight:
206.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

mp

168-170 °C (lit.)

SMILES string

CC1OC[C@H]2O[C@H](O)[C@H](O)[C@@H](O)[C@@H]2O1

InChI

1S/C8H14O6/c1-3-12-2-4-7(13-3)5(9)6(10)8(11)14-4/h3-11H,2H2,1H3/t3?,4-,5-,6-,7-,8+/m1/s1

InChI key

VZPBLPQAMPVTFO-NKWOADHPSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Janoshazi et al.
The Journal of membrane biology, 132(2), 167-178 (1993-03-01)
The kinetics of the initial phases of D-glucose binding to the glucose transport protein (GLUT1) of the human red cell can be followed by stopped-flow measurements of the time course of tryptophan (trp) fluorescence enhancement. A number of control experiments
F R Gorga et al.
Biochemistry, 21(8), 1905-1908 (1982-04-13)
The effect of ligands on the tryptophan fluorescence of the purified monosaccharide transporter from human erythrocytes has been investigated. Cytochalasin B, D-glucose, and ethylideneglucose quench the fluorescence of the protein at longer wavelengths by 17%, 13%, and 8%, respectively. Propyl
J R Appleman et al.
Biochemistry, 28(20), 8221-8227 (1989-10-03)
There is considerable evidence that the mechanism of glucose transport by the transporter of human erythrocytes is one in which the transporter oscillates between two conformations, To and Ti. Each conformer possesses a single glucose binding site that in vivo
H Ishihara et al.
Biochemical and biophysical research communications, 176(2), 922-930 (1991-04-30)
GLUT1 glucose transporter cDNA was modified to introduce a single amino acid substitution of aspartic acid for asparagine 415, which is conserved among all facilitative glucose transporter isoforms. Although a significant amount of the mutated transporter was expressed into plasma
Y Oka et al.
The Journal of biological chemistry, 259(13), 8125-8133 (1984-07-10)
Irradiation of intact rat adipocytes with high intensity ultraviolet light in the presence of 0.5 microM [3H] cytochalasin B results in the labeling of Mr 43,000 and 46,000 proteins that reside in the plasma membrane fraction. In contrast to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service