E20004
Ethyl 2-oxocyclohexanecarboxylate
95%
Synonym(s):
Ethyl cyclohexanone-2-carboxylate, 2-(Ethoxycarbonyl)cyclohexanone, Ethyl-2-oxocyclohexancarboxylat
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(O=)C6H9CO2C2H5
CAS Number:
Molecular Weight:
170.21
Beilstein:
608356
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
95%
form
liquid
refractive index
n20/D 1.477 (lit.)
bp
106 °C/11 mmHg (lit.)
density
1.064 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C1CCCCC1=O
InChI
1S/C9H14O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h7H,2-6H2,1H3
InChI key
FGSGHBPKHFDJOP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
204.8 °F - closed cup
Flash Point(C)
96 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service