Key Documents
DWKW217931
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity (1,250 mL), white polypropylene cap, (PTFE faced LDPE liner), case of 6 ea
Synonym(s):
W217931
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
(PTFE faced LDPE liner)
amber soda-lime glass bottle
white polypropylene cap
Agency
suitable for EPA 1613
feature
with cap
with cap
packaging
case of 6 ea
manufacturer/tradename
WHEATON® W217931
bottle diam. × H
(108 mm) (192 mm)
capacity
(1,250 mL)
fitting
70-400 neck
Features and Benefits
• Ideal for storage of light sensitive samples or dry powders• Easy removal of contents• HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification• Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)• Amber, USP Type III soda-lime glass
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.