Key Documents
DWK885300-0000
KIMBLE® Dounce tissue grinder set
capacity (0.5 mL)
Synonym(s):
KIMBLE tissue grinder, Tissue homogenizer
About This Item
UNSPSC Code:
41101703
NACRES:
NB.74
material
borosilicate glass
packaging
case of 1 ea
manufacturer/tradename
Kimble® 885300-0000
capacity
(0.5 mL)
General description
Dounce All-Glass tissue grinders designed for cellular work in which the nucleus remains intact after homogenization
  • All-glass
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completly interchangeable
  • Manufactured from 33 expansion, low extractaable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Features and Benefits
  • Manufactured from borosilicate glass 3.3
  • Designed primarily for cellular work where the nucleus remains intact after homogenization.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)