Key Documents
DWK73800-13415
KIMBLE® Solid Cap
black phenolic cap, cap for 13-425
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
black phenolic cap
sterility
non-sterile
feature
class A
packaging
case of 1000 ea
manufacturer/tradename
Kimble® 73800-13415
fitting
cap for 13-425
General description
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirement
Features and Benefits
• Autoclavable• White rubber liners• Excellent for general laboratory use• Identified in ASTM Specification E982, Class A requirement
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences