Key Documents
DWK45066C-15
KIMBLE® Solid Cap
black phenolic cap (with PTFE Liner), cap for 15-415
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
PTFE/rubber liner
black phenolic cap (with PTFE Liner)
sterility
non-sterile
feature
class B
packaging
case of 300 ea
manufacturer/tradename
Kimble® 45066C-15
fitting
cap for 15-415
Features and Benefits
• Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving• PTFE faced/general purpose white rubber liners are highly resistant to chemical effects• Excellent for general laboratory use• Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences