Key Documents
DWK258522102
DURAN® Filter funnel
borosilicate glass 3.3, volume 125  mL, porosity 1, autoclavable, pack of 1 ea
Synonym(s):
258522102
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
material
borosilicate glass 3.3
feature
autoclavable
packaging
pack of 1 ea
manufacturer/tradename
DURAN® 258522102
volume
125  mL
pore size
1 porosity
General description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Büchner funnels are cylindrical with an integral sintered disc. Without additional filter paper, only coarse filtration is possible. Büchner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
Features and Benefits
• Manufactured from 3.3. expansion borosilicate glass• Funnels have an integral sintered glass filtration disc• Comprehensive range of funnel sizes and sinter porosities (grades) are available• The use of filter paper is not required• Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid• Filter funnels can be used with a filtering flask via a conical rubber seal (GUKO)• Fired-on, highly durable white print
Legal Information
DURAN is a registered trademark of DWK Life Sciences