Key Documents
DWK218062454
DURAN® graduated amber laboratory bottle
with cap, round amber glass bottle, bottle capacity (100 mL), non-sterile
Synonym(s):
218062454, DURAN® Amber Bottle, Graduated Glass Bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
round amber glass bottle
sterility
non-sterile
feature
with cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 218062454
bottle capacity
(100 mL)
General description
Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties.
Very durable and chemically resistant amber colour. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print. Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Legal Information
DURAN is a registered trademark of DWK Life Sciences