Skip to Content
MilliporeSigma
All Photos(1)

Documents

D39002

Sigma-Aldrich

1,2-Dibromobenzene

98%

Synonym(s):

o-Dibromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Br2
CAS Number:
Molecular Weight:
235.90
Beilstein:
970241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.2 (vs air)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.611 (lit.)

bp

224 °C (lit.)

mp

4-6 °C (lit.)

density

1.956 g/mL at 25 °C (lit.)

SMILES string

Brc1ccccc1Br

InChI

1S/C6H4Br2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

WQONPSCCEXUXTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dibromobenzene, also known as o-Dibromobenzene, is an aromatic halogenated hydrocarbon commonly used as a precursor in various organic synthesis reactions based on the intermediate formation of benzynes.

Application


  • Selective Covalent Capture of Collagen Triple Helices with Minimal Protecting Group Strategy: This study details the use of Rink Amide MBHA resin for capturing collagen triple helices, highlighting its efficiency in solid-phase synthesis (JDH Le Tracy Yu, 2020).

  • Solid phase peptide synthesis: new resin and new protecting group: Introduction of a novel resin, Fmoc-Rink-Amide PEG Octagel, showcasing its application in peptide synthesis (S Ramkisson, 2018).

  • Development of a novel, automated, robotic system for rapid, high-throughput, parallel, solid-phase peptide synthesis: This paper discusses the utilization of Rink Amide MBHA resin in developing an automated system for peptide synthesis (K Kiss et al., 2023).

  • Synthesis of Peptoids Containing Multiple Nhtrp and Ntrp Residues: A Comparative Study of Resin, Cleavage Conditions and Submonomer Protection: The study compares different resins including Rink Amide MBHA, analyzing their efficacy in peptoid synthesis (A Lone et al., 2020).



It is also used as a key reactant in the synthesis of:
  • diborylbenzenes via palladium-catalyzed borylation of bromobenzenes
  • o,o′-tri- and -tetrasubstituted biphenyls via aryne cross-coupling reaction

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselectivity in the Aryne Cross-Coupling of Aryllithiums with Functionalized 1, 2-Dibromobenzenes
Vincent D, et al.
European Journal of Organic Chemistry (2011)
Palladium-Catalyzed Double Cross-Coupling Reactions of Organodimetallic Reagents Leading to Polycyclic Aromatic Hydrocarbons
Masaki S, et al.
European Journal of Organic Chemistry, 2013, 8069-8081 (2013)
Efficient and Complementary Methods Offering Access to Synthetically Valuable 1, 2-Dibromobenzenes
Vincent D, et al.
European Journal of Organic Chemistry (2011)
G Shakila et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 449-455 (2011-11-25)
The FT-IR and FT-Raman spectra of the compound 1,2-dibromobenzene have been recorded in the region 4000-100cm(-1). The vibrational analysis has been made using HF and DFT (B3LYP and LSDA) level of theory by employing 6-31 +G (d, p) and 6-311
Holger F Bettinger et al.
The Journal of organic chemistry, 72(25), 9750-9752 (2007-11-08)
The one-step high-yield synthesis of 1,2-bis(trimethylsilyl)benzene from 1,2-dibromobenzene using tert-butyllithium and trimethylsilyltriflate is reported. A mechanistic investigation shows that 1,2-dilithiobenzene is not an intermediate in this reaction; the coexistence of trimethylsilyltriflate and tert-butyllithium at very low temperatures allows a sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service