Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D31729

Sigma-Aldrich

Dibenzosuberenol

Synonym(s):

5H-Dibenzo[a,d]cyclohepten-5-ol, NSC 84190

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

122.5-124 °C (lit.)

SMILES string

OC1c2ccccc2C=Cc3ccccc13

InChI

1S/C15H12O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-10,15-16H

InChI key

SRIISEYIFDTFRZ-UHFFFAOYSA-N

Application

Reactant involved in:
  • Oxidation with tert-butylhydroperoxide in water for synthesis of carboxylic acids and ketones
  • Synthesis of tachykinin neurokinin-1 receptor antagonists
  • Pauson-Khand reaction mechanism studies
  • Mapping the active site of aziridination catalysts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service