Skip to Content
MilliporeSigma

D26024

3,4-Diaminotoluene

97%

Synonym(s):

4-Methyl-o-phenylenediamine, 3,4-Toluenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
207-826-2
Beilstein/REAXYS Number:
507965
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

solid

bp

155-156 °C/18 mmHg (lit.)

mp

87-89 °C (lit.)

SMILES string

Cc1ccc(N)c(N)c1

InChI

1S/C7H10N2/c1-5-2-3-6(8)7(9)4-5/h2-4H,8-9H2,1H3

InChI key

DGRGLKZMKWPMOH-UHFFFAOYSA-N



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library