Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D196401

Sigma-Aldrich

2,7-Dinitrofluorene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N2O4
CAS Number:
Molecular Weight:
256.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

SMILES string

[O-][N+](=O)c1ccc-2c(Cc3cc(ccc-23)[N+]([O-])=O)c1

InChI

1S/C13H8N2O4/c16-14(17)10-1-3-12-8(6-10)5-9-7-11(15(18)19)2-4-13(9)12/h1-4,6-7H,5H2

InChI key

IHZCVUBSTYOFSJ-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Malejka-Giganti et al.
Carcinogenesis, 20(10), 2017-2023 (1999-10-03)
Nitrofluorene compounds are environmental pollutants chiefly from incomplete combustion. This study examined carcinogenicities after one intramammary injection of 2-nitrofluorene (2-NF), 2, 7-dinitrofluorene (2,7-diNF) or dimethyl sulfoxide (DMSO) (solvent control) to 30-day-old and of 2-NF, 9-OH-2-NF, 9-oxo-2-NF, 2,7-diNF, 9-oxo-2,7-diNF, 2,5-dinitrofluorene, 9-oxo-2,4,7-trinitrofluorene
Clare L Ritter et al.
Chemical research in toxicology, 15(4), 536-544 (2002-04-16)
Nitrofluorenes are mutagenic and carcinogenic environmental pollutants arising chiefly from combustion of fossil fuels. Nitro aromatic compounds undergo nitroreduction to N-hydroxy arylamines that bind to DNA directly or after O-esterification. This study analyzes the DNA binding and adducts from the
C L Ritter et al.
Chemical research in toxicology, 13(8), 793-800 (2000-08-24)
Nitrofluorenes and C-9-oxidized nitrofluorenes are widespread environmental genotoxins which may be relevant for breast cancer on the basis of their carcinogenicities, particularly of 2, 7-dinitrofluorene (2,7-diNF), for the rat mammary gland. Since their metabolism to active carcinogens may involve nitroreduction
L Möller et al.
Carcinogenesis, 14(12), 2627-2632 (1993-12-01)
The urinary mutagenicity (unconjugated forms) after administration of 2,7-dinitrofluorene (2,7-dNF) orally or i.p. was lower compared to 2-nitrofluorene (NF) administration. When partial hepatectomy (PH) was performed before i.p. administration, both substances had higher excretion of mutagens in which 2,7-dNF increased
Danuta Malejka-Giganti et al.
International journal of cancer, 122(9), 1958-1965 (2008-01-10)
Environmental pollution with nitroaromatic compounds may pose health hazards. We have examined the tumorigenicity in female Sprague-Dawley rats of 2,7-dinitrofluorene (2,7-diNF) and 9-oxo-2,7-diNF administered by intraperitoneal (i.p.) and oral routes at 10 micromol/kg body weight, 3 times per week for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service