Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

D174009

Sigma-Aldrich

2,3-Dimethylphenol

98%

Synonym(s):

3-Hydroxyl-o-xylene, vic.-o-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1906267
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.23 (vs air)

Assay

98%

form

crystals

bp

217 °C (lit.)

mp

70-73 °C (lit.)

SMILES string

Cc1cccc(O)c1C

InChI

1S/C8H10O/c1-6-4-3-5-8(9)7(6)2/h3-5,9H,1-2H3

InChI key

QWBBPBRQALCEIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Removal of phenolic compounds from aqueous solutions using poly (styrene-co-divinylbenzene) functionalized with aminophosphonic acid: This research explores the effectiveness of specific polymer adsorbents in removing 2,3-dimethylphenol from water (CM Davidescu et al., 2014).
  • Adsorption of phenol or phenol derivatives onto styrene-1%(15%) divinylbenzene polymeric adsorbents functionalized with aminopropyl (benzyl) phosphonic: Focuses on the removal of 2,3-dimethylphenol from aqueous solutions, highlighting the efficiency of newly developed adsorbents (R Ardelean et al., 2016).
  • Carpe diene! Europium-catalyzed and rearrangements of aryl-pentadienyl ethers: Describes a study where 2,3-dimethylphenol was used to test the catalytic properties of Europium in organic reactions (M Kaiser et al., 2023).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Baggi et al.
Applied and environmental microbiology, 53(9), 2129-2132 (1987-09-01)
A Pseudomonas stutzeri strain capable of growing on o-xylene was isolated from enrichment cultures. The organism grew on 2,3- and 3,4-dimethylphenol but not on 2-methylbenzyl alcohol, o-tolualdehyde, or o-toluate. P. stutzeri was not able to utilize m-xylene, p-xylene, or 1,2,4-trimethylbenzene
A E Holliday et al.
Journal of chemical ecology, 38(3), 278-286 (2012-03-07)
The defensive secretion of the ground beetle Chlaenius cordicollis is predominantly 3-methylphenol. Adult C. cordicollis were collected in Pennsylvania and Manitoba and induced to discharge defensive secretion in a vial. The headspace was sampled by solid phase microextraction, and samples
Jee Hee Suh et al.
Chemical & pharmaceutical bulletin, 63(8), 573-578 (2015-06-05)
We describe the synthesis and biological evaluation of N-aryl-5-aryloxazol-2-amine derivatives that are able to inhibit 5-lipoxygenase (5-LOX), a key enzyme of leukotriene synthesis, for the treatment of inflammation-related diseases including asthma and rheumatoid arthritis. A novel structural moiety containing oxazole
Fangfang Zeng et al.
Insect biochemistry and molecular biology, 113, 103213-103213 (2019-08-24)
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1
Zulin Zhang et al.
Journal of separation science, 37(24), 3699-3705 (2014-10-02)
A simple and rapid method was developed for the simultaneous analysis of nine different pesticides in water samples by gas chromatography with mass spectrometry. A number of parameters that may affect the recovery of pesticides, such as the type of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service