Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D144800

Sigma-Aldrich

3-(Dimethylamino)propiophenone hydrochloride

99%

Synonym(s):

β-(Dimethylamino)propiophenone hydrochloride, β-DAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2CH2N(CH3)2 · HCl
CAS Number:
Molecular Weight:
213.70
Beilstein:
3568398
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

mp

150-155 °C (lit.)

SMILES string

Cl.CN(C)CCC(=O)c1ccccc1

InChI

1S/C11H15NO.ClH/c1-12(2)9-8-11(13)10-6-4-3-5-7-10;/h3-7H,8-9H2,1-2H3;1H

InChI key

DKNDBIIKSJWQFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service