Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D133809

Sigma-Aldrich

3,4-Dimethoxycinnamic acid, predominantly trans

99%

Synonym(s):

Caffeic acid dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

181-183 °C (lit.)

SMILES string

COc1ccc(\C=C\C(O)=O)cc1OC

InChI

1S/C11H12O4/c1-14-9-5-3-8(4-6-11(12)13)7-10(9)15-2/h3-7H,1-2H3,(H,12,13)/b6-4+

InChI key

HJBWJAPEBGSQPR-GQCTYLIASA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Chemical constituents of Veronicastrum sibiricum (L.) Pennell].
B Zhou et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 17(1), 35-36 (1992-01-01)
Tracy L Farrell et al.
Molecular nutrition & food research, 56(9), 1413-1423 (2012-08-07)
This study reports the 24 h human plasma pharmacokinetics of 3,4-dimethoxycinnamic acid (dimethoxycinnamic acid) after consumption of coffee, and the membrane transport characteristics of certain dimethoxycinnamic acid derivatives, as present in coffee. Eight healthy human volunteers consumed a low-polyphenol diet
Thiago Nogueira et al.
Electrophoresis, 28(19), 3570-3574 (2007-09-05)
A method based on the formation of pi-complexes with chlorogenate-like species was proposed for the determination of caffeine in regular (nondecaffeinated) and decaffeinated coffee. Both caffeate and 3,4-dimethoxycinnamate were able to transform caffeine--a neutral species in aqueous solutions--into an anionic
Maher M El-Domiaty et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 64(1-2), 11-19 (2009-03-28)
A new natural compound, named 6-O-(3",4"-dimethoxycinnamoyl) catalpol, was isolated from the defatted alcoholic extract of the flowering parts of Buddleja asiatica Lour. (family Scrophulariaceae). Other separated known compounds included steroids (beta-sitosterol, stigmasterol, stigmasterol-O-glucoside, beta-sitosterol-O-glucoside), iridoid glucosides (methyl catalpol, catalpol, aucubin)
A Sethuraman et al.
Applied microbiology and biotechnology, 52(5), 689-697 (1999-11-26)
Production of ligninolytic enzymes and degradation of 14C-ring labeled synthetic lignin by the white-rot fungus Cyathus stercoreus ATCC 36910 were determined under a variety of conditions. The highest mineralization rate for 14C dehydrogenative polymerizates (DHP; 38% 14CO2 after 30 days)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service