Skip to Content
MilliporeSigma
All Photos(1)

Documents

D122408

Sigma-Aldrich

4,4′-Diiodobiphenyl

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4C6H4I
CAS Number:
Molecular Weight:
406.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

powder

mp

201-204 °C (lit.)

SMILES string

Ic1ccc(cc1)-c2ccc(I)cc2

InChI

1S/C12H8I2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H

InChI key

GPYDMVZCPRONLW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiko Yamada-Okabe et al.
Toxicology letters, 155(1), 127-133 (2004-12-09)
Previously, we demonstrated that some endocrine disrupting chemicals affected thyroid hormone receptor (TR)-mediated gene expression in HeLaTR cells that stably expressed the human TRalpha1. To examine whether widely used brominated flame retardants and pesticides affect TR-mediated gene expression, those with
Dan-Dan Kong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(27), 9895-9904 (2015-05-28)
The synthesis and characterization of a series of biphenyl-derived binuclear ruthenium complexes with terminal {RuCl(CO)(PMe3)3} moieties and different structural arrangements of the phenyl rings are reported. Electrochemical studies revealed that the two metal centers of the binuclear ruthenium complexes interact
L van Bree et al.
Journal of biochemical toxicology, 5(1), 57-63 (1990-01-01)
The effects of pretreatment with symmetrically dihalogenated biphenyls (DXBs, X-F, Cl(C), Br(B) and I) on rat liver drug metabolism enzymes were investigated. 4,4'-DFB, -DCB, and -DBB as well as 2,2'-DFB appeared to be inducers of microsomal cytochrome P-450-linked monoxygenases (N-demethylases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service