D100900
N,N′-Diethylthiourea
98%
Synonym(s):
1,3-Diethyl-2-thiourea, 1,3-Diethylthiourea, N,N′-Diethylthiocarbamide, N,N′-Diethylthiourea
About This Item
Recommended Products
Assay
98%
form
crystals
mp
76-78 °C (lit.)
SMILES string
CCNC(=S)NCC
InChI
1S/C5H12N2S/c1-3-6-5(8)7-4-2/h3-4H2,1-2H3,(H2,6,7,8)
InChI key
FLVIGYVXZHLUHP-UHFFFAOYSA-N
Gene Information
human ... EPHX2(2053)
mouse ... Ephx2(13850)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- 3,5-Diethyltetrahydro-4-thioxo-1,3,5-triazine-1(2H)-propanoic acid by condensation reaction with formaldehyde and β-alanine.[1]
- Methyl 3-ethyl-2-(ethylimino)-3,4-dihydro-4-oxo-2H-1,3-thiazine-6-carboxylate by one-pot reaction with dimethyl acetylenedicarboxylate in the presence of triphenylphosphine catalyst.[2]
- 2-(Ethylimino)-3-ethyl-thiazolidin-4-one by cyclization reaction with 2-chloroacetic acid.[3]
- 3-Ethyl-2-(ethylimino)-4-oxo-5-thiazolidineacetamide by condensation reaction with maleimides via Michael-type reaction.[4]
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1B
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
366.8 °F
Flash Point(C)
186 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service