Key Documents
CLS814040
Pyrex® heavy-duty graduated centrifuge tube, beaded rim
capacity 40 mL
Synonym(s):
Corning® 8140
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
material
conical bottom tube
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 8140-40
O.D. × L
28 mm × 135 mm
capacity
40 mL
General description
The 12 mL size is graduated upward from thip in 0.1 mL divisions. The 40 mL size is graduated in 0.5 mL divisions to 10 mL, and from 10 to 40 mL in 1 mL divisions.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.