Key Documents
CLS808050
Pyrex® centrifuge tubes with beaded rim and white enamel graduations
capacity 50 mL
Synonym(s):
Corning® 8080
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
material
conical bottom tube
sterility
non-sterile
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 8080-50
O.D. × L
28 mm × 135 mm
capacity
50 mL
General description
Formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches, chips, and cracks. Always use with correct sleeve or holder.
Other Notes
15 mL sizes have 0.1 mL graduations upward from tip; 50 mL size has 0.5 mL graduations upward from tip to 10 mL and 1 mL graduations from 10 to 50 mL.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.