Key Documents
CLS76248
Pyrex® HDPE stopper, improved form
stopper joint: 8
Synonym(s):
flask stopper, flask stoppers, stoppers, Corning® 7624
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
Joint
joint: stopper8
manufacturer/tradename
Corning 7624-8
General description
Blue and white stoppers with closed bottoms, made of linear high-density polyethylene. Improved design fully protects the neck in the event of accidental tipping.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)