CLS7082516X
Pyrex® VISTA™ culture tube, screw-cap
diam. × L 16 mm × 125 mm
Synonym(s):
culture tubes, test tubes, test tubes glass, Corning® 70825
About This Item
Recommended Products
material
borosilicate glass
cap (Rubber liner)
round bottom tube
sterility
non-sterile
packaging
case of 200
manufacturer/tradename
Corning 70825-16X
diam. × L
16 mm × 125 mm
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service