Key Documents
CLS4100250
Pyrex® flat-bottom boiling flask, short neck with 24/40 joint
capacity 250 mL
Synonym(s):
boiling flask, flask glass, flat bottom flasks, Corning® 4100
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
short
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 12
manufacturer/tradename
Corning 4100-250
capacity
250 mL
neck size
(1)
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)