Key Documents
CLS4060250
Pyrex® Florence flat-bottom flask
capacity 250 mL
Synonym(s):
bottom flask, flat flask, florence flask, Corning® 4060
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
stopper size
5
sterility
non-sterile
feature
closure type Tooled mouth
long neck, tooled mouth
packaging
case of 24
manufacturer/tradename
Corning 4060-250
capacity
250 mL
neck size
(1)
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.