Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3950

Corning® PYREX® borosilicate glass fiber (BGF) filter discs 8 μm pore size

borosilicate glass fiber (BGF), pore size 8 μm

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
40161507
NACRES:
NB.43

Product Name

Pyrex® fiber glass wool, borosilicate glass fiber (BGF), pore size 8 μm

material

borosilicate glass fiber (BGF)

Agency

suitable for EPA 1613

sterility

non-sterile

packaging

box of 454 g 1box = 2 x 227g bags

manufacturer/tradename

Corning 3950

diam. × L

2 in. × 22 ft

pore size

8 μm

General description

This fiber glass is made from fine glass fibers with diameters controlled to approximately 0.008 mm. This makes the glass pliable and easy to use. Manufactured from Code No. 9989 glass, this fiber glass contains minimal heavy metals and is suitable for use as a filtering medium.

Supplied as a 2 in. diameter x 22 ft long piece.

Features and Benefits

  • Pyrex® fiber glass wool, pore size 8 μm is made of borosilicate glass fiber (BGF).
  • Designed for use in heating mantles, hot plate heating, oven and microwave.
  • May be autoclaved.
  • Not applicable for vacuum or pressure applications.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service