MilliporeSigma
CLS33601LFO
Pyrex® general purpose distilling apparatus, Pyrex stopper, ST joint
stopper and flask only, capacity 1,000 mL
Synonym(s):
Corning® 3360
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.43
UNSPSC Code:
41121800
Capacity:
1,000 mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint stopper27
manufacturer/tradename
Corning 3360-1LFO
capacity
1,000 mL
General description
A general purpose batch distillation assembly with the facility or addition via the Pyrex stoppered neck. Ideally suited for the batch production of high purity distilled water. The flask has a flat bottom with a side delivery tube, terminating in a joint. Condenser tubulations are 10mm O.D.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
CLS33601LFO-1EA04061833884027