Key Documents
CLS3294050M
Pyrex® Gooch crucible, high form, with fritted disc
capacity 50 mL, porosity 10-15 μm
Synonym(s):
Corning® 32940
About This Item
UNSPSC Code:
41121812
manufacturer/tradename
Corning 32940-50M
capacity
50 mL
disc diam.
40 mm
pore size
10-15 μm porosity
General description
Particularly adapted to analytical work where precipitates are dried to constant weight at 110°C. Also suitable for higher temperatures, in which case is advisable to heat in an electric furnace.
Crucibles should not be subjected to sudden temperature changes.
In order to avoid strain, they should not be removed from the furnace until the temperature has dropped to 250°C.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)