Key Documents
CLS2400300
Pyrex® Liebig condenser with top and bottom joints
jacket L 300 mm, joint: ST/NS 24/40
Synonym(s):
Corning® 2400
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
manufacturer/tradename
Corning 2400-300
availability
not available in EU (Not available in EU countries)
jacket L
300 mm
General description
With drip-tip. Large-capacity water jacket cools efficiently.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)