Key Documents
CLS1760125
Pyrex® tall form gas washing bottle, with ST stopper
capacity 125 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
feature
cap
packaging
case of 1 ea
pack of 1 ea
manufacturer/tradename
Corning 1760-125
capacity
125 mL
diam. × H
41 mm × 245 mm
Legal Information
PYREX is a registered trademark of Corning, Inc.