Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS000251

Sigma-Aldrich

1,1-Diphenylhydrazine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
Molecular Weight:
184.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

NN(c1ccccc1)c2ccccc2

InChI

1S/C12H12N2/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,13H2

InChI key

YHYKLKNNBYLTQY-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anuradha Singh et al.
Journal of labelled compounds & radiopharmaceuticals, 61(4), 386-390 (2017-12-24)
Metabolism, environmental fate, and low concentration food residue studies would be facilitated by the use of radiolabeled test articles that can be readily quantified within complex matrices. However, radiochemical approaches for such studies require high specific activities to allow analytical
Rushika Patel et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(2), 126-141 (2019-10-01)
On-line detection of aromatic hydrocarbon pollutants in aqueous environments can be achieved by biosensing strains having fusion of gene responsible for pollutant sensing protein with a reporter gene. Regulatory proteins TbuT, HbpR and PhnR are such proteins for recognizing one-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service