Key Documents
CAD9928B
Animal feeding needle, sterile, disposable, PTFE w/SS bite protector
size 18G, L × diam. 1.5 in. × 2 mm , ball
Synonym(s):
Feeding Needle with Bite Protector, PTFE Feeding Needle
About This Item
UNSPSC Code:
42142609
NACRES:
NB.03
size
18G
packaging
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
1.5 in. × 2 mm , ball
General description
  • Flexible PTFE tube, covered at the hub end with type 304 stainless steel preventing tube damage due to biting during use
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box