Key Documents
CAD6185
Adapter, FLL to Fistula tip (plated brass)
FLL to fistula tip (plated brass)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.03
packaging
pack of 10 ea
manufacturer/tradename
Cadence Science 6185
General description
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.