Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C99604

Sigma-Aldrich

Cycloheptylamine

99%

Synonym(s):

Aminocycloheptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H13NH2
CAS Number:
Molecular Weight:
113.20
Beilstein:
1847543
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

54 °C/11 mmHg (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

NC1CCCCCC1

InChI

1S/C7H15N/c8-7-5-3-1-2-4-6-7/h7H,1-6,8H2

InChI key

VXVVUHQULXCUPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F

Flash Point(C)

42 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neelu Kaila et al.
Bioorganic & medicinal chemistry, 15(19), 6425-6442 (2007-08-01)
We have previously reported the discovery and initial SAR of the [1,7]naphthyridine-3-carbonitriles and quinoline-3-carbonitriles as Tumor Progression Loci-2 (Tpl2) kinase inhibitors. In this paper, we report new SAR efforts which have led to the identification of 4-alkylamino-[1,7]naphthyridine-3-carbonitriles. These compounds show
Julia Greiser et al.
Dalton transactions (Cambridge, England : 2003), 47(27), 9000-9007 (2018-06-21)
We report the isolation of a new type of 1,4-diazepan-6-amine (DAZA)-based ligand. Condensation of aldehydes with DAZA gives a novel class of 1,5-diazabicyclo[3.2.1]octanes in nearly quantitative yields. Subsequent reductive cleavage of these bicyclic aminal species with sodium borohydride selectively leads
Stelios Eleftheratos et al.
Bioorganic & medicinal chemistry letters, 20(14), 4182-4187 (2010-06-24)
Interaction of aminoadamantanes with the influenza A virus M2 proton channel was analyzed by docking simulations of a series of synthetic aminoadamantane derivatives, of differing binding affinity, into the crystal structure of the transmembrane (M2TM) pore. The pore blocking model
Tsung-Chi Chen et al.
Rapid communications in mass spectrometry : RCM, 25(21), 3274-3280 (2011-10-19)
The discontinuous atmospheric pressure interface (DAPI) has been developed to allow a direct transfer of ions from atmosphere into an ion trap mass spectrometer with minimum pumping capability. Air is introduced into the trap with ions and used as a
Oxygen-18 studies on the oxidative deamination mechanism of alicyclic primary amines in rabbit liver microsomes.
H Kurebayashi et al.
Archives of biochemistry and biophysics, 215(2), 433-443 (1982-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service