Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8561

Sigma-Aldrich

Cadaverine dihydrochloride

~98%

Synonym(s):

1,5-Diaminopentane dihydrochloride, 1,5-Pentanediamine dihydrochloride, Pentamethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)5NH2 · 2HCl
CAS Number:
Molecular Weight:
175.10
Beilstein:
3611329
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

~98%

form

powder

mp

260-262 °C (lit.)

SMILES string

Cl.Cl.NCCCCCN

InChI

1S/C5H14N2.2ClH/c6-4-2-1-3-5-7;;/h1-7H2;2*1H

InChI key

FGNLEIGUMSBZQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cadaverine dihydrochloride can be used as a:
  • Starting material in the synthesis of pentamethylene diisocyanate (PDI) via phosgenation and dechlorination.
  • Linker in the preparation of trinuclear palladium complexes by using trans-palladin as the starting material.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangqun Lan et al.
Journal of food science and technology, 57(12), 4414-4423 (2020-10-23)
Nattokinase activity (NK), biogenic amine content and sensory properties of natto are of great significance to consumers, which are affected by strains and fermentation methods. In this study, changes in the pH, biogenic amine and free amino nitrogen (FAN) contents, NK
J Wang et al.
Journal of animal science, 93(4), 1679-1688 (2015-05-29)
Polyamines are necessary for normal integrity and the restitution after injury of the gastrointestinal epithelium. The objective of this study was to investigate the effects of oral administration of putrescine and proline during the suckling period on epithelial restitution after
Elvira S Plakidi et al.
Foods (Basel, Switzerland), 9(5) (2020-05-14)
Monitoring of biogenic amines in food is important for quality control, in terms of freshness evaluation and even more for food safety. A novel and cost-effective method was developed and validated for the determination of the main biogenic amines: histamine
Thomas Boehm et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 69(9), 937-950 (2020-06-04)
To measure diamine oxidase (DAO) activity with high sensitivity in complex matrices like plasma or tissue extracts radioactive putrescine or horseradish peroxidase (HRP)/hydrogen peroxide (H2O2) coupling must be used. The use of radioactive material should be avoided and HRP/H2O2 coupling
Julia Torres et al.
Organic & biomolecular chemistry, 13(27), 7500-7512 (2015-06-13)
Within all the eukaryotic cells myo-inositol phosphates (InsPs) are an important group of biomolecules that are potentially related to signaling functions. The most abundant member of this family in nature is InsP6 (phytate, L(12-) in its fully deprotonated form). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service