Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

C70606

Sigma-Aldrich

4-Chlororesorcinol

98%

Synonym(s):

1,3-Dihydroxy-4-chlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OH)2
CAS Number:
Molecular Weight:
144.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

147 °C/18 mmHg (lit.)

mp

106-108 °C (lit.)

SMILES string

Oc1ccc(Cl)c(O)c1

InChI

1S/C6H5ClO2/c7-5-2-1-4(8)3-6(5)9/h1-3,8-9H

InChI key

JQVAPEJNIZULEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guadalupe Albarrán et al.
Environmental science and pollution research international, 27(18), 22855-22864 (2020-04-24)
Radiation-induced degradation of chlorobenzene was conducted at 0.1, 0.4, 0.5, 0.7, and 1.0 mmol/dm3 concentrations in aerated environment and at 1.0 mmol/dm3 in oxygen-free and N2O-saturated solutions. The results demonstrated that the elimination of chloride is important when the solution is oxygen
Burcu Bekdeşer et al.
Journal of agricultural and food chemistry, 62(46), 11109-11115 (2014-11-05)
Hypochlorous acid (HOCl) is an important reactive oxygen species (ROS) and non-radical and is taking part in physiological processes concerned with the defense of the organism, but there has been limited information regarding its scavenging by polyphenols. This study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service