Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

C70401

Sigma-Aldrich

2-Chloroquinoline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6ClN
CAS Number:
Molecular Weight:
163.60
Beilstein:
112561
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

266-267 °C (lit.)

mp

34-37 °C (lit.)

density

1.23 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc2ccccc2n1

InChI

1S/C9H6ClN/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

OFUFXTHGZWIDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Fetzner et al.
FEMS microbiology letters, 112(2), 151-157 (1993-09-01)
Resting cells of Pseudomonas putida strain 86 were grown on quinoline transformed 2-chloroquinoline to 2-chloro-cis-7,8-dihydro-7,8-dihydroxyquinoline which was not converted further. 7,8-Dioxygenating activity was present when the enzymes of quinoline catabolism were induced. Quinoline-grown cells of strain 86 treated simultaneously with
K Shiva Kumar et al.
Bioorganic & medicinal chemistry, 20(7), 2199-2207 (2012-03-06)
A number of 2-(1H-indol-3-yl)quinoline-3-carbonitrile derivatives were synthesized via AlCl(3)-mediated C-C bond forming reaction between 2-chloroquinoline-3-carbonitrile and various indoles. The methodology does not require any N-protection of the indoles employed and provided the corresponding products in good yields. The molecular structure
D R Boyd et al.
Organic & biomolecular chemistry, 8(5), 1081-1090 (2010-02-19)
A series of enantiopure 2,2'-bipyridines have been synthesised from the corresponding cis-dihydrodiol metabolites of 2-chloroquinolines. Several of the resulting hydroxylated 2,2'-bipyridines were found to be useful chiral ligands for the asymmetric aminolysis of meso-epoxides leading to the formation of enantioenriched
Amol A Nagargoje et al.
Chemistry & biodiversity, 17(2), e1900624-e1900624 (2019-12-22)
In search for new fungicidal and free radical scavenging agents, we synthesized a focused library of 2-chloroquinoline based monocarbonyl analogs of curcumin (MACs). The synthesized MACs were evaluated for in vitro antifungal and antioxidant activity. The antifungal activity was evaluated against
Yubin Ji et al.
Biomedical chromatography : BMC, 34(8), e4845-e4845 (2020-04-09)
An ultra-high performance liquid chromatography tandem mass spectrometry method was developed for determination of homocysteine (HCY) in human plasma. The HCY was derivatized with 2-chloro-1-methylquinolinium tetrafluoroborate and isolated using solid-phase extraction. Derivatization, isolation and detection procedures were optimized. Satisfactory linearity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service