Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C54201

Sigma-Aldrich

1-(Chloromethyl)naphthalene

90%

Synonym(s):

1-Naphthylmethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2Cl
CAS Number:
Molecular Weight:
176.64
Beilstein:
636885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

solid

refractive index

n20/D 1.635 (lit.)

bp

167-169 °C/25 mmHg (lit.)

mp

32 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

ClCc1cccc2ccccc12

InChI

1S/C11H9Cl/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2

InChI key

XMWGTKZEDLCVIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

remainder 2-(chloromethyl)naphthalene

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service