Key Documents
BR92736
BRAND® Erlenmeyer flask with beaded rim and graduation, narrow mouth
Synonym(s):
Erlenmeyer flasks, Flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom
Joint
joint neck34 mm
sterility
non-sterile
feature
without cap
packaging
pack of 10 ea
manufacturer/tradename
BRAND 92736
General description
The BRAND® Erlenmeyer Flask with beaded rim and graduations is a high-quality laboratory vessel designed for versatile use in chemical and biological applications. Made from Boro 3.3 glass, this flask meets DIN ISO 1773 standards, ensuring durability and reliability in various laboratory environments.
Features and Benefits
  • Good mechanical resistance
  • Easy to clean
  • Diverse applications
Other Notes
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)